7-Chloro-4-chromanone 97%

Supplier: Ambeed
Warning

A424916-100MG A424916-10G A424916-1G A424916-250MG
76694-984EA 22.28 USD
76694-984 76694-986 76694-988 76694-990
7-Chloro-4-chromanone 97%
7-Chloro-4-chromanone

Precaución: for Research Use Only

Formula: C₉H₇ClO₂
MW: 182.6000 g/mol
MDL Number: MFCD09744054
CAS Number: 18385-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR