Tyrphostin A23 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3,4-Dihydroxybenzylidene)malononitrile

A423886-10MG A423886-1MG A423886-5MG
76694-974EA 83.82 USD
76694-974 76694-976 76694-978
Tyrphostin A23 98%
Tyrphostin A23

Precaución: for Research Use Only

Formula: C₁₀H₆N₂O₂
MW: 186.1 g/mol
MDL Number: MFCD00133899
CAS Number: 118409-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR