8-Pentadecanone 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Diheptyl ketone, Caprylone
A423071-25G
A423071-5G
76694-970EA
177.18
USD
76694-970
76694-972
8-Pentadecanone 99%
8-Pentadecanone
Precaución: for Research Use Only
Formula:
C₁₅H₃₀O MW: 226.4 g/mol |
MDL Number:
MFCD00009555 CAS Number: 818-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...