Butyl oleate 98%

Supplier: Ambeed
Warning

Synonyms: Butyl (Z)-9-octadecenoate, Oleic acid butyl ester

A418841-100G A418841-500G
76694-962EA 5.31 USD
76694-962 76694-964
Butyl oleate 98%
Butyl oleate

Precaución: for Research Use Only

Formula: C₂₂H₄₂O₂
MW: 338.57 g/mol
MDL Number: MFCD00053819
CAS Number: 142-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR