7-(Dimethylamino)-4-(trifluoromethyl)coumarin 98%

Supplier: Ambeed
Warning

Synonyms: 7-(Dimethylamino)-4-(trifluoromethyl)-2H-chromen-2-one

A418606-1G A418606-5G
76694-958EA 28.65 USD
76694-958 76694-960
7-(Dimethylamino)-4-(trifluoromethyl)coumarin 98%
7-(Dimethylamino)-4-(trifluoromethyl)coumarin

Precaución: for Research Use Only

Formula: C₁₂H₁₀F₃NO₂
MW: 257.2085 g/mol
MDL Number: MFCD00041867
CAS Number: 53518-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR