Benzeneseleninic anhydride 98%

Supplier: Ambeed
Danger

A418092-5G A418092-1G
76694-956EA 163 USD
76694-956 77594-294
Benzeneseleninic anhydride 98%
Benzeneseleninic anhydride

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₃Se₂
MDL Number: MFCD00001991
CAS Number: 17697-12-0
UN: 3283
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR