Allyltriethylammonium bromide 97%

Supplier: Ambeed
Warning

A417677-25G A417677-5G
76694-948EA 34 USD
76694-948 76694-950
Allyltriethylammonium bromide 97%
Allyltriethylammonium bromide

Precaución: for Research Use Only

Formula: CH₂=CHCH₂N(CH₂CH₃)₃Br
MDL Number: MFCD00051865
CAS Number: 29443-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR