2-(3-Methoxypropoxy)-4-((R)-2-(bromomethyl)-3-methylbutyl)-1-methoxybenzene 96%

Supplier: Ambeed
Warning

A415473-1G A415473-250MG A415473-5G
76694-934EA 76.39 USD
76694-934 76694-936 76694-938
2-(3-Methoxypropoxy)-4-((R)-2-(bromomethyl)-3-methylbutyl)-1-methoxybenzene 96%
2-(3-Methoxypropoxy)-4-((R)-2-(bromomethyl)-3-methylbutyl)-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₇H₂₇BrO₃
MW: 359.30 g/mol
MDL Number: MFCD10566721
CAS Number: 172900-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR