AC-Ser(tBu)-OH 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Acetamido-3-(tert-butoxy)propanoic acid, N-Acetyl-O-tert-butyl-L-serine

A415452-1G A415452-25G A415452-5G
76694-928EA 13.8 USD
76694-928 76694-930 76694-932
AC-Ser(tBu)-OH 98%
AC-Ser(tBu)-OH

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
MDL Number: MFCD00236770
CAS Number: 77285-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR