Benzotriazol-1-ylacetic acid 96%

Supplier: Ambeed
Warning

Synonyms: 2-(1H-Benzo[d][1,2,3]triazol-1-yl)acetic acid

A415433-1G A415433-250MG A415433-25G A415433-5G
76694-920EA 46.68 USD
76694-920 76694-922 76694-924 76694-926
Benzotriazol-1-ylacetic acid 96%
Benzotriazol-1-ylacetic acid

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
MDL Number: MFCD00553342
CAS Number: 4144-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR