Cyclobutanol 98%

Supplier: Ambeed
Danger

A415172-1G A415172-5G A415172-25G A415172-10G
76694-916EA 9.55 USD
76694-916 76694-918 77551-246 77568-840
Cyclobutanol 98%
Cyclobutanol

Precaución: For research use only.

Formula: C₄H₈O
MW: 72.11 g/mol
MDL Number: MFCD00001318
CAS Number: 2919-23-5
UN: 1987
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR