7-Benzyl-5,6,7,8-tetrahydro-3H-pyrido[3,4-d]pyrimidin-4-one 97%

Supplier: Ambeed
Warning

Synonyms: 7-Benzyl-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidin-4(3H)-one

A414868-1G A414868-250MG A414868-5G
76694-910EA 51.99 USD
76694-910 76694-912 76694-914
7-Benzyl-5,6,7,8-tetrahydro-3H-pyrido[3,4-d]pyrimidin-4-one 97%
7-Benzyl-5,6,7,8-tetrahydro-3H-pyrido[3,4-d]pyrimidin-4-one

Precaución: for Research Use Only

Formula: C₁₄H₁₅N₃O
MW: 241.3 g/mol
MDL Number: MFCD09030048
CAS Number: 62458-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR