cis-1,2-Cyclohexanediol 98%

Supplier: Ambeed
Warning

Synonyms: cis-Cyclohexane-1,2-diol

A413282-1G A413282-250MG A413282-5G
76694-892EA 50 USD
76694-892 76694-894 76694-896
cis-1,2-Cyclohexanediol 98%
cis-1,2-Cyclohexanediol

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 120
MDL Number: MFCD00064944
CAS Number: 1792-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR