Ethyl-3,5-dichlorobenzoate 98%

Supplier: Ambeed
Warning

A413158-100G A413158-10G A413158-25G A413158-5G
76694-884EA 29 USD
76694-884 76694-886 76694-888 76694-890
Ethyl-3,5-dichlorobenzoate 98%
Ethyl-3,5-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₂
MDL Number: MFCD00221467
CAS Number: 91085-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR