Ethyl-4-acetyl-5-oxohexanoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4,4-Diacetylbutyrate, Ethyl 4-acetyl-5-oxohexanoate, 4,4-Diacetylbutyric Acid Ethyl Ester, 4-Acetyl-5-oxohexanoic Acid Ethyl Ester

A412801-1G A412801-25G A412801-5G
76694-870EA 12 USD
76694-870 76694-872 76694-874
Ethyl-4-acetyl-5-oxohexanoate 98%
Ethyl-4-acetyl-5-oxohexanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₄
MDL Number: MFCD00009211
CAS Number: 2832-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR