9-Methyladenine 95%

Supplier: Ambeed
Warning

Synonyms: 9-Methyl-9H-purin-6-amine

A411838-100MG A411838-1G A411838-250MG
76694-854EA 54.11 USD
76694-854 76694-856 77601-794
9-Methyladenine 95%
9-Methyladenine

Precaución: for Research Use Only

Formula: C₆H₇N₅
MW: 149.16 g/mol
MDL Number: MFCD00047232
CAS Number: 700-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR