D-(-)-Tartaric acid 98%

Supplier: Ambeed
Warning

A411816-100G A411816-25G A411816-500G A411816-1000G
76694-848EA 7.43 USD
76694-848 76694-850 76694-852 77594-260
D-(-)-Tartaric acid 98%
D-(-)-Tartaric acid

Precaución: for Research Use Only

Formula: C₄H₆O₆
MDL Number: MFCD00004238
CAS Number: 147-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR