4,5-Diphenyl-1,3-thiazol-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 4,5-Diphenylthiazol-2-amine

A411540-100MG A411540-1G A411540-250MG A411540-5G
76694-840EA 20.16 USD
76694-840 76694-842 76694-844 76694-846
4,5-Diphenyl-1,3-thiazol-2-amine 97%
4,5-Diphenyl-1,3-thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂S
MW: 252.34 g/mol
MDL Number: MFCD00438803
CAS Number: 6318-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR