4,4,13,13-Tetraethoxy-3,14-dioxa-8,9-dithia-4,13-disilahexadecane 12.5-15.5% S

Supplier: Ambeed
Warning

A410681-100G A410681-10G A410681-1G A410681-25G A410681-5G
76694-822EA 23.34 USD
76694-822 76694-824 76694-826 76694-828 76694-830
4,4,13,13-Tetraethoxy-3,14-dioxa-8,9-dithia-4,13-disilahexadecane 12.5-15.5% S
4,4,13,13-Tetraethoxy-3,14-dioxa-8,9-dithia-4,13-disilahexadecane

Precaución: for Research Use Only

Formula: C₁₈H₄₂O₆S₂Si₂
MW: 474.82 g/mol
MDL Number: MFCD05663903
CAS Number: 56706-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR