7-(Diethylamino)-4-(trifluoromethyl)coumarin 97%

Supplier: Ambeed
Warning

Synonyms: Coumarin 481, 7-(Diethylamino)-4-(trifluoromethyl)-2H-chromen-2-one

A407743-1G A407743-250MG
76694-810EA 122.01 USD
76694-810 76694-812
7-(Diethylamino)-4-(trifluoromethyl)coumarin 97%
7-(Diethylamino)-4-(trifluoromethyl)coumarin

Precaución: for Research Use Only

Formula: C₁₄H₁₄F₃NO₂
MW: 285.27 g/mol
MDL Number: MFCD00057313
CAS Number: 41934-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR