ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form) 99%

Supplier: Ambeed
Warning

A407582-1G A407582-250MG A407582-5G
76694-804EA 84 USD
76694-804 76694-806 76694-808
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form) 99%
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form)

Precaución: for Research Use Only

Formula: C₂₁H₂₇N₇NaO₁₇P₃
MDL Number: MFCD00036973
CAS Number: 1184-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR