Chloro(indenyl)bis(triphenylphosphine)ruthenium(II),dichloromethane adduct 98%

Supplier: Ambeed
Warning

Synonyms: Chloro(indenyl)bis(triphenylphosphine)ruthenium(II)

A405048-100MG A405048-1G A405048-250MG A405048-5G
76694-794EA 10 USD
76694-794 76694-796 76694-798 76694-800
Chloro(indenyl)bis(triphenylphosphine)ruthenium(II),dichloromethane adduct 98%
Chloro(indenyl)bis(triphenylphosphine)ruthenium(II),dichloromethane adduct

Precaución: for Research Use Only

Formula: C₄₅H₃₇ClP₂Ru
MW: 776.2500 g/mol
MDL Number: MFCD04113601
CAS Number: 99897-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR