Boc-L-HomoSer(Bzl)-OH 95%

Supplier: Ambeed
Warning

Synonyms: Boc-Hse(Bzl)-OH, Boc-HomoSer(Bzl)-OH

A404818-1G A404818-250MG
76694-790EA 27 USD
76694-790 76694-792
Boc-L-HomoSer(Bzl)-OH 95%
Boc-L-HomoSer(Bzl)-OH

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₅
MDL Number: MFCD00270497
CAS Number: 59408-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR