Bosentan monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: 4-tert-Butyl-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-2-(pyrimidin-2-yl)pyrimidin-4-yl]benzenesulphonamide, 4-(tert-Butyl)-N-(6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-[2,2'-bipyrimidin]-4-yl)benzenesulphonamide hydrate, Bosentan hydrate, 4-(tert-Butyl)-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)[2,2'-bipyrimidin]-4-yl]benzenesulphonamide

A404664-100MG A404664-1MG A404664-50MG A404664-10MG A404664-1G A404664-250MG A404664-5G
76694-784EA 20.16 USD
76694-784 76694-786 76694-788 77601-774 77601-776 77601-778 77601-780
Bosentan monohydrate 98%
Bosentan monohydrate

Precaución: for Research Use Only

Formula: C₂₇H₂₉N₅O₆S·H₂O
MW: 551.62 g/mol
MDL Number: MFCD09751188
CAS Number: 157212-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR