Adenosine-3',5'-cyclic monophosphate sodium salt 95%

Supplier: Ambeed
Warning

Synonyms: 3′,5′-Cyclic AMP sodium salt, 3',5'-Cyclic AMP Sodium Salt, Adenosine 3',5'-Cyclic Monophosphate Sodium Salt Hydrate, 3',5'-cAMP Na, (4aR,6R,7R,7aS)-6-(6-Amino-9H-purin-9-yl)-2,7-dihydroxytetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinine 2-oxide, sodium salt, cAMP-Na, 3',5'-Cyclic AMP Sodium Salt Hydrate, 3',5'-cAMP Na Hydrate

A404489-1G A404489-250MG A404489-10G
76694-780EA 40 USD
76694-780 76694-782 77878-149
Adenosine-3',5'-cyclic monophosphate sodium salt 95%
Adenosine-3',5'-cyclic monophosphate sodium salt

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₅NaO₆P
MW: 351.19 g/mol
MDL Number: MFCD00069736
CAS Number: 37839-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR