Fuchsin (acid) BS

Supplier: Ambeed
Warning

Synonyms: CI 42685

A402817-100G A402817-25G A402817-5G A402817-500G
76694-766EA 88 USD
76694-766 76694-768 76694-770 77626-816
Fuchsin (acid) BS
Fuchsin (acid)

Precaución: for Research Use Only

Formula: C₂₀H₁₇N₃Na₂O₉S₃
MDL Number: MFCD00013286
CAS Number: 3244-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR