2-Bromo-9-phenyl-9H-fluoren-9-ol 95%

Supplier: Ambeed
Warning

A400915-1G A400915-250MG A400915-5G
76694-756EA 78 USD
76694-756 76694-758 76694-760
2-Bromo-9-phenyl-9H-fluoren-9-ol 95%
2-Bromo-9-phenyl-9H-fluoren-9-ol

Precaución: for Research Use Only

Formula: C₁₉H₁₃BrO
MDL Number: MFCD09953787
CAS Number: 736928-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR