7-Amino-heptan-1-ol 97%

Supplier: Ambeed
Danger

A399260-100MG A399260-1G A399260-250MG A399260-5G A399260-10G A399260-25G
76694-750EA 7 USD
76694-750 76694-752 76694-754 77572-234 77878-137 77878-138
7-Amino-heptan-1-ol 97%
7-Amino-heptan-1-ol

Precaución: for Research Use Only

Formula: C₇H₁₇NO
MDL Number: MFCD02094314
CAS Number: 19243-04-0
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR