Bis(phenylthio)methane 97%

Supplier: Ambeed
Warning

A395356-1G A395356-25G A395356-5G
76694-726EA 6.37 USD
76694-726 76694-728 76694-730
Bis(phenylthio)methane 97%
Bis(phenylthio)methane

Precaución: for Research Use Only

Formula: C₁₃H₁₂S₂
MW: 232.37 g/mol
MDL Number: MFCD00003067
CAS Number: 3561-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR