Z-Val-Ome 98%

Supplier: Ambeed
Warning

A394562-100MG A394562-250MG
76694-720EA 25 USD
76694-720 76694-722
Z-Val-Ome 98%
Z-Val-Ome

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MDL Number: MFCD00672538
CAS Number: 24210-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR