3-(4-Fluorophenyl)acrylonitrile 99%

Supplier: Ambeed
Warning

A394011-1G A394011-250MG A394011-5G
76694-714EA 44.56 USD
76694-714 76694-716 76694-718
3-(4-Fluorophenyl)acrylonitrile 99%
3-(4-Fluorophenyl)acrylonitrile

Precaución: for Research Use Only

Formula: C₉H₆FN
MW: 147.14 g/mol
MDL Number: MFCD03093768
CAS Number: 24654-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR