Methyl-3,4-dimethylbenzoate 98%

Supplier: Ambeed
Warning

A391889-1G A391889-25G A391889-5G
76694-686EA 10 USD
76694-686 76694-688 76694-690
Methyl-3,4-dimethylbenzoate 98%
Methyl 3,4-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MDL Number: MFCD00219861
CAS Number: 38404-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR