Adipamide 98%

Supplier: Ambeed
Warning

A391480-100G A391480-1G A391480-25G A391480-5G
76694-658EA 371.32 USD
76694-658 76694-660 76694-662 76694-664
Adipamide 98%
Adipamide

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O₂
MW: 144.17 g/mol
MDL Number: MFCD00008045
CAS Number: 628-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR