(S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol ≥97%, ee 99%

Supplier: Ambeed
Danger

A390911-1G A390911-250MG A390911-5G
76694-652EA 56 USD
76694-652 76694-654 76694-656
(S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol ≥97%, ee 99%
(S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol

Precaución: for Research Use Only

Formula: C₂₀H₁₂Br₂O₂
MDL Number: MFCD03093629
CAS Number: 119707-74-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR