4-Nitro-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: H-Phe(4-NO2)-OH, (S)-4-Nitrophenylalanine, (S)-2-Amino-3-(4-nitrophenyl)propanoic acid

A390472-100G A390472-10G A390472-25G A390472-500G A390472-5G
76694-640EA 48.81 USD
76694-640 76694-642 76694-644 76694-646 76694-648
4-Nitro-L-phenylalanine 97%
4-Nitro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₄
MW: 210.19 g/mol
MDL Number: MFCD00051221
CAS Number: 949-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR