2-(Allyloxy)benzaldehyde 96%

Supplier: Ambeed
Warning

Synonyms: Salicylaldehyde Allyl Ether, 2-Allyloxybenzaldehyde

A390119-10G A390119-1G A390119-5G A390119-25G
76694-630EA 49 USD
76694-630 76694-632 76694-634 77878-125
2-(Allyloxy)benzaldehyde 96%
2-(Allyloxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MDL Number: MFCD00014130
CAS Number: 28752-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR