1,2,3,4-Hetrahydroacridin-9-amine hydrochloride monohydrate 97%

Supplier: Ambeed
Danger

Synonyms: Tetrahydroaminacrine hydrochloride monohydrate, 1,2,3,4-Hetrahydroacridin-9-amine hydrochloride hydrate, Tacrine hydrochloride monohydrate, 1,2,3,4-Hetrahydroacridin-9-amine hydrochloride monohydrate, THA hydrochloride monohydrate

A389995-1G A389995-250MG A389995-5G
76694-624EA 65 USD
76694-624 76694-626 76694-628
1,2,3,4-Hetrahydroacridin-9-amine hydrochloride monohydrate 97%
1,2,3,4-Hetrahydroacridin-9-amine hydrochloride monohydrate

Precaución: for Research Use Only

Formula: C₁₃H₁₇ClN₂O
MDL Number: MFCD00150067
CAS Number: 7149-50-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR