Calcium 2-hydroxypropanoate pentahydrate 98%

Supplier: Ambeed
Warning

A388042-1000G A388042-100G A388042-25G A388042-500G
76694-604EA 23 USD
76694-604 76694-606 76694-608 76694-610
Calcium 2-hydroxypropanoate pentahydrate 98%
Calcium 2-hydroxypropanoate pentahydrate

Precaución: for Research Use Only

Formula: C₆H₂₀CaO₁₁
MDL Number: MFCD00150987
CAS Number: 5743-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR