2,5-Dioxopyrrolidin-1-yl octanoate 97%

Supplier: Ambeed
Warning

A386850-10G A386850-250MG A386850-5G A386850-1G
76694-598EA 98 USD
76694-598 76694-600 76694-602 77594-178
2,5-Dioxopyrrolidin-1-yl octanoate 97%
2,5-Dioxopyrrolidin-1-yl octanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₄
MDL Number: MFCD00050404
CAS Number: 14464-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR