Clinofibrate 97%

Supplier: Ambeed
Warning

A385904-10MG A385904-1MG A385904-50MG A385904-5MG
76694-572EA 8 USD
76694-572 76694-574 76694-576 76694-578
Clinofibrate 97%
Clinofibrate

Precaución: for Research Use Only

Formula: C₂₈H₃₆O₆
MDL Number: MFCD00865769
CAS Number: 30299-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR