5-Chloro-m-xylene 97%

Supplier: Ambeed
Warning

A385541-1G A385541-25G A385541-5G
76694-560EA 8.49 USD
76694-560 76694-562 76694-564
5-Chloro-m-xylene 97%
5-Chloro-m-xylene

Precaución: for Research Use Only

Formula: C₈H₉Cl
MDL Number: MFCD00060273
CAS Number: 556-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR