2-(Piperidin-2-yl)benzo[d]thiazole 97%

Supplier: Ambeed
Warning

A384777-100MG A384777-250MG
76694-544EA 53 USD
76694-544 77626-758
2-(Piperidin-2-yl)benzo[d]thiazole 97%
2-(Piperidin-2-yl)benzo[d]thiazole

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂S
MW: 218.31 g/mol
MDL Number: MFCD05190089
CAS Number: 51785-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR