Allyl-4-methylbenzenesulfonate 98%

Supplier: Ambeed
Warning

A383575-100MG A383575-1G A383575-250MG A383575-5G
76694-512EA 6 USD
76694-512 76694-514 76694-516 76694-518
Allyl-4-methylbenzenesulfonate 98%
Allyl-4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃S
MDL Number: MFCD00093777
CAS Number: 4873-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR