8-Bromo-4-chloro-2-(trifluoromethyl)quinoline 97%

Supplier: Ambeed
Warning

Synonyms: 8-Bromo-4-chloro-2-(trifluoromethyl)-1-azanaphthalene

A382925-250MG A382925-5G
76694-502EA 38.2 USD
76694-502 76694-504
8-Bromo-4-chloro-2-(trifluoromethyl)quinoline 97%
8-Bromo-4-chloro-2-(trifluoromethyl)quinoline

Precaución: for Research Use Only

Formula: C₁₀H₄BrClF₃N
MW: 310.498 g/mol
MDL Number: MFCD00174293
CAS Number: 655235-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR