3-Chlorobenzo[b]thiophene-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A382919-5G A382919-100G A382919-1G A382919-25G
76694-500EA 55 USD
76694-500 77878-102 77878-103 77878-104
3-Chlorobenzo[b]thiophene-2-carboxylic acid 98%
3-Chlorobenzo[b]thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅ClO₂S
MDL Number: MFCD00014579
CAS Number: 21211-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR