Bis(7-oxabicyclo[4.1.0]heptan-3-ylmethyl) adipate 95%
Supplier: Ambeed
|
Warning
|
A381806-10G
A381806-5G
A381806-25G
A381806-500G
76694-496EA
7
USD
76694-496
76694-498
76841-058
77626-746
Bis(7-oxabicyclo[4.1.0]heptan-3-ylmethyl) adipate 95%
Bis(7-oxabicyclo[4.1.0]heptan-3-ylmethyl) adipate
Precaución: for Research Use Only
Formula:
C₂₀H₃₀O₆ |
MDL Number:
MFCD00191811 CAS Number: 3130-19-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...