N-(α)-Boc-D-tryptophanol 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Tryptophanol, Boc-D-Trp-OL

A381455-1G A381455-25G A381455-5G
76694-490EA 19.1 USD
76694-490 76694-492 76694-494
N-(α)-Boc-D-tryptophanol 97%
N-(alpha)-Boc-D-tryptophanol

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₂O₃
MW: 290.3 g/mol
MDL Number: MFCD00270221
CAS Number: 158932-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR