2-(4-Fluorophenyl)-2-propanol 96%

Supplier: Ambeed
Warning

Synonyms: alpha,alpha-Dimethyl-4-fluorobenzyl alcohol, 2-(4-Fluorophenyl)propan-2-ol

A381448-1G A381448-25G A381448-5G
76694-484EA 15.92 USD
76694-484 76694-486 76694-488
2-(4-Fluorophenyl)-2-propanol 96%
2-(4-Fluorophenyl)-2-propanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO
MW: 154.18 g/mol
MDL Number: MFCD00059919
CAS Number: 402-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR