Diethyl pyrazole-3,5-dicarboxylate 98%

Supplier: Ambeed
Warning

A379492-100G A379492-10G A379492-1G A379492-25G A379492-5G
76694-440EA 122 USD
76694-440 76694-442 76694-444 76694-446 76694-448
Diethyl pyrazole-3,5-dicarboxylate 98%
Diethyl pyrazole-3,5-dicarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₄
MDL Number: MFCD00152167
CAS Number: 37687-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR