2-[1,1'-Biphenyl]-4-yl-1-ethanol 95%

Supplier: Ambeed
Warning

A377799-100MG A377799-1G A377799-250MG A377799-5G
76694-428EA 6 USD
76694-428 76694-430 76694-432 76694-434 76694-436
2-[1,1'-Biphenyl]-4-yl-1-ethanol 95%
2-[1,1'-Biphenyl]-4-yl-1-ethanol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O
MDL Number: MFCD09926182
CAS Number: 37729-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR